EMAIL THIS PAGE TO A FRIEND
Diabetologia
Nicotinamide mononucleotide protects against pro-inflammatory cytokine-mediated impairment of mouse islet function.
PMID 21901281
Abstract
Nicotinamide phosphoribosyltransferase (NAMPT), the rate-limiting enzyme for NAD(+) biosynthesis, exists as intracellular NAMPT (iNAMPT) and extracellular NAMPT (eNAMPT). eNAMPT, secreted from adipose tissue, promotes insulin secretion. Administration of nicotinamide mononucleotide (NMN), a product of the eNAMPT reaction, corrects impaired islet function in Nampt ( +/- ) mice. One of its potential targets is the NAD(+)-dependent deacetylase sirtuin 1. We hypothesised that altered NAMPT activity might contribute to the suppression of islet function associated with inflammation, and aimed to determine whether NMN could improve cytokine-mediated islet dysfunction. Acute effects of NMN on cytokine-mediated islet dysfunction were examined in islets incubated with TNFα and IL1β, and in mice fed a fructose-rich diet (FRD) for 16 weeks. Changes in iNAMPT, eNAMPT and inflammation levels were determined in FRD-fed mice. FRD-fed mice displayed markedly lower levels of circulating eNAMPT, with impaired insulin secretion and raised islet expression of Il1b. NMN administration lowered Il1b expression and restored suppressed insulin secretion in FRD-fed mice. NMN also restored insulin secretion in islets cultured with pro-inflammatory cytokines. The changes in islet function corresponded with changes in key markers of islet function and differentiation. The anti-inflammatory effects of NMN were partially blocked by inhibition of sirtuin 1. Chronic fructose feeding causes severe islet dysfunction in mice. Onset of beta cell failure in FRD-fed mice may occur via lowered secretion of eNAMPT, leading to increased islet inflammation and impaired beta cell function. Administration of exogenous NMN to FRD-fed mice corrects inflammation-induced islet dysfunction. Modulation of this pathway may be an attractive target for amelioration of islet dysfunction associated with inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3501
β-Nicotinamide mononucleotide, ≥95% (HPLC)
C11H15N2O8P
72340
Nicotinamide, ≥99.5% (HPLC)
C6H6N2O