EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Catalytic syntheses of N-heterocyclic ynones and ynediones by in situ activation of carboxylic acids with oxalyl chloride.
PMID 21910193