EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Discovery of a pharmacologically active antagonist of the two-pore-domain potassium channel K2P9.1 (TASK-3).
PMID 21916012
Abstract
TWIK-related acid-sensitive K(+) (K(2P) 9.1, TASK-3) ion channels have the capacity to regulate the activity of neuronal pathways by influencing the resting membrane potential of neurons on which they are expressed. The central nervous system (CNS) expression of these channels suggests potential roles in neurologic disorders, and it is believed that the development of TASK-3 antagonists could lead to the therapeutic treatment of a number of neurological conditions. While a therapeutic potential for TASK-3 channel modulation exists, there are only a few documented examples of potent and selective small-molecule channel blockers. Herein, we describe the discovery and lead optimization efforts for a novel series of TASK-3 channel antagonists based on a 5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine high-throughput screening lead from which a subseries of potent and selective inhibitors were identified. One compound was profiled in detail with respect to its physical properties and demonstrated pharmacological target engagement as indicated by its ability to modulate sleep architecture in rodent electroencephalogram (EEG) telemetry models.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1326
PK-THPP, ≥98% (HPLC)
C29H32N4O2