EMAIL THIS PAGE TO A FRIEND
Dermatologic clinics
Dapsone in the management of autoimmune bullous diseases.
PMID 21924998
Abstract
Dapsone is used in the treatment of autoimmune bullous diseases (AIBD), a group of disorders resulting from autoimmunity directed against basement membrane and/or intercellular adhesion molecules on cutaneous and mucosal surfaces. This review summarizes the limited published data evaluating dapsone as a therapy for AIBD.