EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and evaluation of the anti parasitic activity of aromatic nitro compounds.
PMID 21940071
Abstract
A series of nitroaromatic compounds was synthesized and evaluated as potential antileishmanial and trypanocidal agents. Five compounds exerted significant anti-leishmanial activity in vitro against promastigotes forms of Leishmania (L.) amazonensis, with IC(50) in the range of 23-59 μmol L(-1), but none were active against amastigotes intracellular forms of Trypanosoma cruzi. In vitro cytotoxicity on the proliferation of human peripheral blood mononuclear cells (PBMC) stimulated with phytohemaglutinin (PHA) was also evaluated. Two compounds, 6 and 7, were found to present a promising anti-leishmanial activity with IC(50) values of 59.5 and 50.6 μM, respectively, without affecting the lymphocyte proliferation in PBMCs (selectivity index of 16.1 and 21.7, respectively), indicating low toxicity to human cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
159549
4-(Bromomethyl)benzoic acid, 97%
C8H7BrO2