EMAIL THIS PAGE TO A FRIEND
Molecular cancer
Transglutaminase 2 as an independent prognostic marker for survival of patients with non-adenocarcinoma subtype of non-small cell lung cancer.
PMID 21943122
Abstract
Expression of transglutaminase 2 (TGase 2) is related to invasion and resistance to chemotherapeutic agents in several cancer cells. However, there has been only limited clinical validation of TGase 2 as an independent prognostic marker in cancer. The significance of TGase 2 expression as an invasive/migratory factor was addressed by in vitro assays employing down-regulation of TGase 2. TGase 2 expression as a prognostic indicator was assessed in 429 Korean patients with early-stage non-small cell lung cancer (NSCLC) by immunohistochemical staining. TGase 2 expression increased the invasive and migratory properties of NSCLC cells in vitro, which might be related to the induction of MMP-9. In the analysis of the immunohistochemical staining, TGase 2 expression in tumors was significantly correlated with recurrence in NSCLC (p = 0.005) or in the non-adenocarcinoma subtype (p = 0.031). Additionally, a multivariate analysis also showed a significant correlation between strong TGase 2 expression and shorter disease-free survival (DFS) in NSCLC (p = 0.029 and HR = 1.554) and in the non-adenocarcinoma subtype (p = 0.030 and HR = 2.184). However, the correlation in the adenocarcinoma subtype was not significant. TGase 2 expression was significantly correlated with recurrence and shorter DFS in NSCLC, especially in the non-adenocarcinoma subtype including squamous cell carcinoma.