EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
One-pot enzymatic production of 2-acetamido-2-deoxy-D-galactose (GalNAc) from 2-acetamido-2-deoxy-D-glucose (GlcNAc).
PMID 21955790
Abstract
2-Acetamido-2-deoxy-D-galactose (GalNAc) is a common monosaccharide found in biologically functional sugar chains, but its availability is often limited due to the lack of abundant natural sources. In order to produce GalNAc from abundantly available sugars, 2-acetamido-2-deoxy-D-glucose (GlcNAc) was converted to GalNAc by a one-pot reaction using three enzymes involved in the galacto-N-biose/lacto-N-biose I pathway of bifidobacteria. Starting the reaction with 600 mM GlcNAc, 170 mM GalNAc was produced at equilibrium in the presence of catalytic amounts of ATP and UDP-Glc under optimized conditions. GalNAc was separated from GlcNAc using water-eluting cation-exchange chromatography with a commonly available cation-exchange resin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0167
Galacto-N-biose
C14H25NO11