EMAIL THIS PAGE TO A FRIEND
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke
[Pivampicillin (Pondocillin) and pivmecillinam (Selexid) can result in carnitine deficiency].
PMID 2195708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0817
Pivmecillinam, ≥98% (HPLC)
C21H33N3O5S