EMAIL THIS PAGE TO A FRIEND
Medicinal research reviews
Polymethylene tetraamines: a novel class of cardioselective M2-antagonists.
PMID 2196404
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M105
Methoctramine hydrate, ≥97% (NMR), solid
C36H62N4O2 · 4HCl · xH2O