EMAIL THIS PAGE TO A FRIEND
Histology and histopathology
PTPIP51, a positive modulator of the MAPK/Erk pathway, is upregulated in glioblastoma and interacts with 14-3-3β and PTP1B in situ.
PMID 21972092
Abstract
Glioblastoma multiforme (GBM) is the most common and most malignant primary brain tumour. Protein tyrosine phosphatase interacting protein 51 (PTPIP51) is an interaction partner of 14-3-3β, which correlates with the grade of malignancy in gliomas. In this study PTPIP51 and its interacting partners 14-3-3β, PTP1B, c-Src, Raf-1 as well as EGFR were investigated in human glioblastoma. Twenty glioblastoma samples were analyzed on transcriptional and translational level by immunohistochemistry, in situ hybridization and RT-PCR. To compare PTPIP51 expression in gliomas of different malignancies, quantitative RT-PCR for grade II astrocytoma and GBM samples was employed. Additionally, we analyzed the correlation between PTPIP51 and 14-3-3β transcription, and checked for in situ interaction between PTPIP51 and 14-3-3β and PTP1B, respectively. PTPIP51 and 14-3-3β mRNA showed a tumour grade dependent upregulation in gliomas. Glioblastoma cells displayed a strong immunoreaction of PTPIP51, which co-localized with 14-3-3β and PTP1B. The duolink proximity ligation assay corroborated a direct in situ interaction of PTPIP51 with both proteins, known to interact with PTPIP51 in vitro. The in vitro interacting partners Raf-1 and c-Src showed a partial co-localization. Besides, immune cells located in capillaries or infiltrating the tumour tissue and endothelial cells of pseudoglomerular vessels revealed a high PTPIP51 expression. The upregulation of PTPIP51 and its connection with the EGFR/MAPK pathway by 14-3-3β via Raf-1 and by PTP1B via c-Src, argue for a functional role of PTPIP51 in the pathogenesis of human glioblastoma.