EMAIL THIS PAGE TO A FRIEND
Biological procedures online
Rapid generation of long tandem DNA repeat arrays by homologous recombination in yeast to study their function in mammalian genomes.
PMID 21982381
Abstract
We describe here a method to rapidly convert any desirable DNA fragment, as small as 100 bp, into long tandem DNA arrays up to 140 kb in size that are inserted into a microbe vector. This method includes rolling-circle phi29 amplification (RCA) of the sequence in vitro and assembly of the RCA products in vivo by homologous recombination in the yeast Saccharomyces cerevisiae. The method was successfully used for a functional analysis of centromeric and pericentromeric repeats and construction of new vehicles for gene delivery to mammalian cells. The method may have general application in elucidating the role of tandem repeats in chromosome organization and dynamics. Each cycle of the protocol takes ~ two weeks to complete.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5501
L-Lysine, ≥98% (TLC)
C6H14N2O2