EMAIL THIS PAGE TO A FRIEND
ChemMedChem
From a library of MAG antagonists to nanomolar CD22 ligands.
PMID 21990163
Abstract
Siglec-2, also known as CD22, is involved in the regulation and survival of B-cells and has been successfully targeted in cell depletion therapies with antibody-based approaches. Sialic acid derivatives, already known to bind with high affinity to myelin-associated glycoprotein (MAG, Siglec-4), were screened for their binding affinity for CD22 by surface plasmon resonance. The best compound identified was further modified with various hydrophobic substituents at the 2-, 5-, and 9-positions of the sialic acid scaffold, leading to nanomolar derivatives, of which ligand 17 b shows the most promising pharmacodynamic and pharmacokinetic profiles. Isothermal titration calorimetry measurements demonstrate that the binding is enthalpy driven. Interestingly, the thermodynamic fingerprints reveal an excellent correlation between gains in enthalpy and compensation by increased entropy costs. Moreover, 17 b exhibits a residence time in the range of a few seconds, clearly prolonged relative to residence times typically observed for carbohydrate-lectin interactions. Finally, initial tests regarding drug-like properties of 17 b demonstrate the required high plasma protein binding yet a lack of oral availability, although its distribution coefficient (log D) is in the required range.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
523526
2,3-Dichlorobenzyl alcohol, 97%
C7H6Cl2O