EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Ligustrazine derivatives. Part 5: design, synthesis and biological evaluation of novel ligustrazinyloxy-cinnamic acid derivatives as potent cardiovascular agents.
PMID 21993151
Abstract
A series of novel ligustrazinyloxy-cinnamic acid derivatives were designed, synthesized and evaluated for their inhibitory effect on adenosine diphosphate (ADP)-induced platelet aggregation in vitro, and also assayed for their protective effect against hydrogen peroxide (H(2)O(2))-induced oxidative damage on ECV-304 cells. Some compounds exhibited high activity in one or both of the assays, of which, compound 2e displayed the highest protective effect on the proliferation of the damaged ECV-304 cells (EC(50) = 0.020 mM), and compound 2f was the most active anti-platelet aggregation agent (EC(50) = 0.054 mM). Structure-activity relationships were briefly discussed.