EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Structural features of the HMG chromosomal proteins and their genes.
PMID 2200521
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SRP2051
HMG1, native from calf thymus, ≥85% (SDS-PAGE)