EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Neuropilin-1 expression in cancer and development.
PMID 22025255
Abstract
Neuropilin (NRP)-1 is a co-receptor for vascular endothelial growth factor (VEGF). Preclinical data suggest that blockade of NRP1 suppresses tumour growth by inhibiting angiogenesis, in addition to directly inhibiting tumour cell proliferation in certain models. A humanized monoclonal antibody to NRP1 is currently being evaluated as a potential anti-cancer therapy in clinical trials. However, the expression of NRP1 in cancer and physiological angiogenesis has yet to be systematically described. Here we characterize the in situ expression of NRP1 in human cancer and during mammalian development. A monoclonal antibody to human NRP1 was generated and validated for immunohistochemistry by western blotting, use of formalin-fixed cell pellets transfected with NRP1, immunofluorescence, and comparison with in situ hybridization. NRP1 expression was assessed in whole sections of 65 primary breast carcinomas, 95 primary colorectal adenocarcinomas, and 90 primary lung carcinomas. An additional 59 human metastases, 16 xenografts, and three genetically engineered mouse tumour models were also evaluated. Immunoreactivity for NRP1 was seen in vessels from normal tissues adjacent to cancer and in 98-100% of carcinomas. Tumour cell expression of NRP1 was also observed in 36% of primary lung carcinomas and 6% of primary breast carcinomas, but no colorectal adenocarcinomas. NRP1 was evaluated in mouse embryos, where expression was limited to the nervous system, endocardium, vascular smooth muscle, and, focally, endothelium on subsets of vessels. Moreover, in a model of VEGF-dependent angiogenesis in the postnatal mouse trachea, blockade of NRP1 signalling resulted in defective angiogenesis and recapitulated the effects of anti-VEGF treatment. These observations confirm NRP1 as a valid anti-angiogenic target in malignancy, and as a potential direct anti-tumour target in a subset of cancers. The data also confirm a role for NRP1 in physiological, VEGF-mediated angiogenesis.
Related Materials