EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Radical cation Diels-Alder cycloadditions by visible light photocatalysis.
PMID 22032252
Abstract
Ruthenium(II) polypyridyl complexes promote the efficient radical cation Diels-Alder cycloaddition of electron-rich dienophiles upon irradiation with visible light. These reactions enable facile [4 + 2] cycloadditions that would be electronically mismatched under thermal conditions. Key to the success of this methodology is the availability of ligand-modified ruthenium complexes that enable rational tuning of the electrochemical properties of the catalyst without significantly perturbing the overall photophysical properties of the system.© 2011 American Chemical Society
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220868
1-Acetoxy-1,3-butadiene, mixture of cis and trans
C6H8O2
Set your institution to read full text papers when available.