PloS one
Impact of genetic variation in SORCS1 on memory retention.
PMID 22046233
Abstract
We previously reported that genetic variants in SORCS1 increase the risk of AD, that over-expression of SorCS1 reduces γ-secretase activity and Aβ levels, and that SorCS1 suppression increases γ-secretase processing of APP and Aβ levels. We now explored the effect of variation in SORCS1 on memory. We explored associations between SORCS1-SNPs and memory retention in the NIA-LOAD case control dataset (162 cases,670 controls) and a cohort of Caribbean Hispanics (549 cases,544 controls) using single marker and haplotype analyses. Three SNPs in intron 1, were associated with memory retention in the NIA-LOAD dataset or the Caribbean Hispanic dataset (rs10884402(A allele:β = -0.15,p = 0.008), rs7078098(C allele:β = 0.18,p = 0.007) and rs950809(C allele:β = 0.17,p = 0.008)) and all three SNPs were significant in a meta-analysis of both datasets (0.002