EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Formation of N-N cross-links in DNA by reaction of radiation-produced DNA base pair diradicals: a DFT study.
PMID 22050209
Abstract
This study employs DFT (density functional theory) to investigate the formation of hydrazine-like (N-N) cross-linked structures between DNA base pair diradicals that are likely to result from the interaction of high linear energy transfer (LET) radiation, such as ion-beam radiation, with DNA. In our calculations, we generated the guanine (G), cytosine (C), adenine (A), and thymine (T) radicals by removing one hydrogen atom from an N-H bond involved in the normal base pairing. The radical species formed are those that naturally result from one-electron oxidation of the bases followed by deprotonation. N-N cross-links between G and C or A and T diradicals were studied using the BHandHLYP, B3LYP, M06, and M06-2X density functionals and 6-31G* basis set. From a comparison to several test cases performed with the G3B3 method, which gives thermodynamically reliable values, we found that calculations employing the BHandHLYP/6-31G* method predict the best estimates of bonding energies for hydrazine-like structures. Our study shows that the N-N cross-link formed between guanine radical and a neutral cytosine is endothermic in nature but can form metastable structures. However, the reactions between two DNA base radicals (diradical) to form several N-N cross-linked structures are found to be highly exothermic in nature. The N-N cross-links formed between various G-C, G-G, and C-C diradicals have binding energies in the range of ca. -54 to -68, -41 to -47, and -67 to -75 kcal/mol, respectively, whereas A-T, A-A, and T-T have binding energies of -80, -60, and -98 kcal/mol, respectively. In all purine-pyrimidine N-N cross-linked structures, the highest occupied molecular orbital (HOMO) is found to be localized on the purine moiety and the lowest unoccupied molecular orbital (LUMO) is on the pyrimidine moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0376
Thymine, ≥99%
C5H6N2O2