PloS one
DNA damage stress and inhibition of Jak2-V617F cause its degradation and synergistically induce apoptosis through activation of GSK3β.
PMID 22087308
Abstract
The cytoplasmic tyrosine kinase Jak2 plays a crucial role in cytokine receptor signaling in hematopoietic cells. The activated Jak2-V617F mutant is present in most cases of BCR/ABL-negative myeloproliferative neoplasms and constitutively activates downstream signals from homodimeric cytokine receptors, such as the erythropoietin receptor (EpoR). Here we examine the effects of DNA damage stress on Jak2 or Jak2-V617F and on induction of apoptosis in hematopoietic cells. Etoposide or doxorubicin dose-dependently decreased the expression level of Jak2 in UT7 or 32D cells expressing EpoR in the absence of Epo and that of exogenously expressed Jak2-V617F in UT7 cells when cotreated with the Jak2 inhibitor JakI-1 or AG490. Studies with pharmacological inhibitors and genetic manipulations further showed that downregulation of the PI3K/Akt pathway leading to the activation of GSK3β may be involved in downregulation of Jak2 or Jak2-V617F as well as in synergistic induction of Bax activation and apoptosis. The downregulation of Jak2 was inhibited by the proteasome inhibitor MG132 or by expression of both of loss-of-function mutants of c-Cbl and Cbl-b, E3 ubiquitin ligases which facilitated ubiquitination of Jak2-V617F when co-expressed in 293T cells. The pan-caspase inhibitor Boc-d-fmk also inhibited the Jak2 downregulation as well as appearance of a 100-kDa fragment that contained the N-terminal portion of Jak2 in response to DNA damage. Together, these data suggest that DNA damage stress with simultaneous inhibition of the kinase activity causes degradation of Jak2 or Jak2-V617F by caspase cleavage and proteasomal degradation through GSK3β activation, which is closely involved in synergistic induction of apoptosis in hematopoietic cells.