PloS one
Regulation of interleukin-10 receptor ubiquitination and stability by beta-TrCP-containing ubiquitin E3 ligase.
PMID 22087322
Abstract
Interleukin-10 (IL-10) initiates potent anti-inflammatory effects via activating its cell surface receptor, composed of IL-10R1 and IL-10R2 subunits. The level of IL-10R1 is a major determinant of the cells' responsiveness to IL-10. Here, via a series of biochemical analyses using 293T cells reconstituted with IL-10R1, we identify the latter as a novel substrate of βTrCP-containing ubiquitin E3 ligase. Within the intracellular tail of IL-10R1, a canonical ((318)DpSGFGpS) and a slightly deviated ((369)DpSGICLQEP) βTrCP recognition motif can additively recruit βTrCP in a phosphorylation-dependent manner. βTrCP recruitment leads to ubiquitination, endocytosis and degradation of IL-10R1, subsequently reducing the cellular responsiveness to IL-10. Our study uncovers a novel negative regulatory mechanism that may potentially affect IL-10 function in target cells under physiological or pathological conditions.