EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Rapid selection of high-affinity binders using ribosome display.
PMID 22094811
Abstract
Ribosome display has proven to be a powerful in vitro selection and evolution method for generating high-affinity binders from libraries of folded proteins. It has been successfully applied to single-chain Fv fragments of antibodies and alternative scaffolds, such as Designed Ankyrin Repeat Proteins (DARPins). High-affinity binders with new target specificity can be obtained from highly diverse DARPin libraries in only a few selection rounds. In this protocol, the selection from the library and the process of affinity maturation and off-rate selection are explained in detail.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41640
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
C2H6OS