EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Towards the prevention of potential aluminum toxic effects and an effective treatment for Alzheimer's disease.
PMID 22099160
Abstract
In 1991, treatment with low dose intramuscular desferrioxamine (DFO), a trivalent chelator that can remove excessive iron and/or aluminum from the body, was reported to slow the progression of Alzheimer's disease (AD) by a factor of two. Twenty years later this promising trial has not been followed up and why this treatment worked still is not clear. In this critical interdisciplinary review, we provide an overview of the complexities of AD and involvement of metal ions, and revisit the neglected DFO trial. We discuss research done by us and others that is helping to explain involvement of metal ion catalyzed production of reactive oxygen species in the pathogenesis of AD, and emerging strategies for inhibition of metal-ion toxicity. Highlighted are insights to be considered in the quests to prevent potentially toxic effects of aluminum toxicity and prevention and intervention in AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266523
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
356859
Aluminum, foil, thickness 0.05 mm, 99.8% trace metals basis
Al
11008
Aluminum, grit, ≥97.0% (complexometric)
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
11009
Aluminum, powder, ≥91% (complexometric)
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
310360
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
11010
Aluminum, powder (fine), ≥93% (complexometric)
Al
424803
Aluminum, disc, 0.5-1.7 mm, ≥99.99% trace metals basis
Al
266590
Aluminum, foil, thickness 2.0 mm, 99.999%
Al
326909
Aluminum, wire, diam. 0.127 mm, ≥99.99%
Al
IRMM523A
Aluminum, IRMM® certified Reference Material, 0.1 mm foil
Al
IRMM523B
Aluminum, IRMM® certified Reference Material, 1.0 mm foil
Al
IRMM523C
Aluminum, IRMM® certified Reference Material, 1.0 mm wire
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al
749036
Aluminum, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.9995% trace metals basis
Al
773964
Aluminum, ingot, L × W 10 mm × 7 mm, 99.99995% trace metals basis
Al
GF81923238
Aluminum, wire reel, 25m, diameter 0.25mm, as drawn, 99.5%
Al