EMAIL THIS PAGE TO A FRIEND
Organic letters
Expanding the azaspiro[3.3]heptane family: synthesis of novel highly functionalized building blocks.
PMID 22111917
Abstract
The preparation of versatile azaspiro[3.3]heptanes carrying multiple exit vectors is disclosed. Expedient synthetic routes enable the straightforward access to these novel modules that are expected to have significance in drug discovery and design.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92459
Trimethylene oxide, purum, ≥97.0% (GC)
C3H6O
T76201
Trimethylene oxide, 97%
C3H6O