EMAIL THIS PAGE TO A FRIEND
Nature
Mutations causing syndromic autism define an axis of synaptic pathophysiology.
PMID 22113615
Abstract
Tuberous sclerosis complex and fragile X syndrome are genetic diseases characterized by intellectual disability and autism. Because both syndromes are caused by mutations in genes that regulate protein synthesis in neurons, it has been hypothesized that excessive protein synthesis is one core pathophysiological mechanism of intellectual disability and autism. Using electrophysiological and biochemical assays of neuronal protein synthesis in the hippocampus of Tsc2(+/-) and Fmr1(-/y) mice, here we show that synaptic dysfunction caused by these mutations actually falls at opposite ends of a physiological spectrum. Synaptic, biochemical and cognitive defects in these mutants are corrected by treatments that modulate metabotropic glutamate receptor 5 in opposite directions, and deficits in the mutants disappear when the mice are bred to carry both mutations. Thus, normal synaptic plasticity and cognition occur within an optimal range of metabotropic glutamate-receptor-mediated protein synthesis, and deviations in either direction can lead to shared behavioural impairments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3689
(S)-3,5-Dihydroxyphenylglycine hydrate, ≥98% (HPLC), powder
C8H9NO4 · xH2O
SML0235
CDPPB, ≥98% (HPLC)
C23H16N4O