EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Mitochondrial electron transport chain complex III is required for antimycin A to inhibit autophagy.
PMID 22118681
Abstract
Autophagy is a cellular lysosome-dependent catabolic mechanism mediating the turnover of intracellular organelles and long-lived proteins. We show that antimycin A, a known inhibitor of mETC complex III, can inhibit autophagy. A structural and functional study shows that four close analogs of antimycin A that have no effect on mitochondria inhibition also do not inhibit autophagy, whereas myxothiazol, another mETC complex III inhibitor with unrelated structure to antimycin A, inhibits autophagy. Additionally, antimycin A and myxothiazol cannot inhibit autophagy in mtDNA-depleted H4 and mtDNA-depleted HeLa cells. These data suggest that antimycin A inhibits autophagy through its inhibitory activity on mETC complex III. Our data suggest that mETC complex III may have a role in mediating autophagy induction.