EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Evaluation of the chiral recognition properties as well as the column performance of four chiral stationary phases based on cellulose (3,5-dimethylphenylcarbamate) by parallel HPLC and SFC.
PMID 22119140
Abstract
The performance of four commercially available cellulose tris(3,5-dimethylphenylcarbamate) based chiral stationary phases (CSPs) was evaluated with parallel high performance liquid chromatography (HPLC) and super critical fluid chromatography (SFC). Retention, enantioselectivity, resolution and efficiency were compared for a set of neutral, basic and acidic compounds having different physico-chemical properties by using different mobile phase conditions. Although the chiral selector is the same in all the four CSPs, a large difference in the ability to retain and resolve enantiomers was observed under the same chromatographic conditions. We believe that this is mainly due to differences in the silica matrix and immobilization techniques used by the different vendors. An extended study of metoprolol and structure analogues gave a deeper understanding of the accessibility of the chiral discriminating interactions and its impact on the resolution of the racemic compounds on the four CSPs studied. Also, a clear difference in enantioselectivity is observed between SFC and LC mode, hydrogen bonding was found to play an important role in the differential binding of the enantiomers to the CSPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K08971227 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 50 mm × 2.1 mm
K08971223 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 150 mm × 2.1 mm
K08971228 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 50 mm × 2.1 mm
K08971224 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 150 mm × 2.1 mm
K08670372 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 50 mm × 4.6 mm
K08670370 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 150 mm × 4.6 mm
K08670617 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 50 mm × 4.6 mm
K08670373 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 150 mm × 4.6 mm
K08670374 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 4.6 mm
K08670614 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 10  mm
K08670615 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 21.2 mm
K08670616 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 30 mm
K08670612 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 150 mm × 4.6 mm
K08670613 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 4.6 mm
K08670609 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 10 mm
K08670610 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 21.2 mm
K08670611 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 30 mm
K08670502 Kromasil® CelluCoat® Chiral Reversed Phase HPLC Column, 3 μm particle size, L × I.D. 50 mm × 4.6 mm
K08670503 Kromasil® CelluCoat® Chiral Reversed Phase HPLC Column, 3 μm particle size, L × I.D. 150 mm × 4.6  mm