PloS one
Partial depletion of natural CD4⁺CD25⁺ regulatory T cells with anti-CD25 antibody does not alter the course of acute influenza A virus infection.
PMID 22125630
Abstract
Foxp3⁺CD4⁺ regulatory T cells represent a T cell subset with well-characterized immunosuppressive effects during immune homeostasis and chronic infections, and there is emerging evidence to suggest these cells temper pulmonary inflammation in response to acute viral infection. Recent studies have demonstrated treatment with PC61 CD25-depleting antibody potentiates inflammation in a murine model of RSV infection, while paradoxically delaying recruitment of CD8⁺ T cells to the site of inflammation. The present study therefore sought to examine the role of these cells in a murine model of acute influenza A virus infection through the administration of PC61 CD25-depleting antibody. PC61 antibody is able to partially deplete CD25⁺Foxp3⁺ regulatory T cells to a comparable degree as seen within previous work examining RSV, however this does not alter influenza A-virus induced mortality, weight loss, viral clearance and cellularity within the lung. Collectively, these data demonstrate that partial depletion of CD4⁺CD25⁺ regulatory T cells with PC61 antibody does not alter the course of influenza A virus infection.