EMAIL THIS PAGE TO A FRIEND
Veterinary journal (London, England : 1997)
Food safety implications of ochratoxin A in animal-derived food products.
PMID 22130462
Abstract
Ochratoxin A (OTA) is an important mycotoxin with potential to reach the human food chain through carry-over of contaminated, mostly cereal-based, feed into animal-derived products. Certain population groups, such as infants and children, are intensive and relatively restricted consumers of some animal-derived products, particularly milk and other dairy products, which may become contaminated with OTA. This review examines the literature on the occurrence of OTA in animal-derived products and discusses the public health and food safety implications of consumption of these products. The risk of OTA contamination of meat, milk, blood and derived products is discussed.