EMAIL THIS PAGE TO A FRIEND
European journal of dermatology : EJD
Identification of a novel U2HR mutation c.14C>T in a Chinese patient with Marie Unna hereditary hypotrichosis.
PMID 22155146
Abstract
Marie Unna hereditary hypotrichosis (MUHH) is an autosomal dominant form of non-syndromic hereditary alopecia. Recently, loss-of-function mutations of an inhibitory upstream open reading frame (ORF) in the human hairless gene (HR), named U2HR, have been identified in some patients with MUHH. We investigated a sporadic Chinese patient with MUHH and identified a novel mutation in U2HR, c.14C>T (p.T5M), which extends the mutation spectrum of U2HR mutations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10289 CompoZr® Knockout ZFN Kit, Human HR (NM_005144)