EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Mechanoinduction of lymph vessel expansion.
PMID 22157817
Abstract
In the mammalian embryo, few mechanical signals have been identified to influence organ development and function. Here, we report that an increase in the volume of interstitial or extracellular fluid mechanically induces growth of an organ system, that is, the lymphatic vasculature. We first demonstrate that lymph vessel expansion in the developing mouse embryo correlates with a peak in interstitial fluid pressure and lymphatic endothelial cell (LEC) elongation. In 'loss-of-fluid' experiments, we then show that aspiration of interstitial fluid reduces the length of LECs, decreases tyrosine phosphorylation of vascular endothelial growth factor receptor-3 (VEGFR3), and inhibits LEC proliferation. Conversely, in 'gain-of-fluid' experiments, increasing the amount of interstitial fluid elongates the LECs, and increases both VEGFR3 phosphorylation and LEC proliferation. Finally, we provide genetic evidence that β1 integrins are required for the proliferative response of LECs to both fluid accumulation and cell stretching and, therefore, are necessary for lymphatic vessel expansion and fluid drainage. Thus, we propose a new and physiologically relevant mode of VEGFR3 activation, which is based on mechanotransduction and is essential for normal development and fluid homeostasis in a mammalian embryo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-1050-M
4G10® Platinum, Anti-Phosphotyrosine Antibody (mouse monoclonal cocktail IgG2b), 4G10 Platinum, Anti-Phosphotyrosine (mouse monoclonal cocktail IgG2b) detects levels of Phosphotyrosine proteins & has been published & validated for use in WB, ICC, ELISA, IHC, IF, Flow & IP.
05-1050
4G10® Platinum, Anti-Phosphotyrosine Antibody (mouse monoclonal cocktail IgG2b), 4G10 Platinum, Anti-Phosphotyrosine (mouse monoclonal cocktail IgG2b) detects levels of Phosphotyrosine proteins & has been published & validated for use in WB, ICC, ELISA, IHC, IF, Flow & IP.
MAB1997
Anti-Integrin β1 Antibody, clone MB1.2, Anti-Integrin β1 Antibody, clone MB1.2 is an antibody against Integrin β1 for use in FC, IH, IP & WB.
MAB1987
Anti-Integrin β1 Antibody, clone P4C10, Detect Integrin β1 using this Anti-Integrin β1 Antibody, clone P4C10 validated for use in ELISA, IP, WB & IC.
CBL1337F
Anti-PECAM-1 Antibody, clone 390, FITC conjugated, This Anti-PECAM-1 Antibody, clone 390, FITC conjugated is validated for use in FC, IH for the detection of PECAM-1.
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
C8487
Anti-Caspase 3, Active antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution