EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Clicking 1,2,4,5-tetrazine and cyclooctynes with tunable reaction rates.
PMID 22159330
Abstract
Substituted tetrazines have been found to undergo facile inverse electron demand Diels-Alder reactions with "tunable" reaction rates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
403555
3,6-Diphenyl-1,2,4,5-tetrazine, 98%
C14H10N4