EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly efficient aerobic oxidative hydroxylation of arylboronic acids: photoredox catalysis using visible light.
PMID 22161996
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480061
2,6-Dimethylphenylboronic acid, ≥95.0%
C8H11BO2
456772
4-Carboxyphenylboronic acid
C7H7BO4