EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Effects of different force fields and temperatures on the structural character of Abeta (12-28) peptide in aqueous solution.
PMID 22174662
Abstract
The aim of this work is to investigate the effects of different force fields and temperatures on the structural character of Aβ (12-28) peptide in aqueous solution. Moreover, the structural character of Aβ (12-28) peptide is compared with other amyloid peptides (such as H1 and α-syn12 peptide). The two independent temperature replica exchange molecular dynamics (T-REMD) simulations were completed by using two different models (OPLS-AA/TIP4P and GROMOS 43A1/SPC). We compared the models by analyzing the distributions of backbone dihedral angles, the secondary structure propensity, the free energy surface and the formation of β-hairpin. The results show that the mostly populated conformation state is random coil for both models. The population of β-hairpin is below 8 percent for both models. However, the peptide modeled by GROMOS 43A1 form β-hairpin with turn located at residues F19-E22, while the peptide modeled by OPLS-AA form β-hairpin with turn located at residues L17-F20.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0034 Amyloid β 12-28 human, ≥98% (HPLC)
C89H135N25O25