EMAIL THIS PAGE TO A FRIEND
Current topics in medicinal chemistry
Melphalan in regional chemotherapy for locally recurrent metastatic melanoma.
PMID 22196271
Abstract
In-transit metastases occur in approximately 3% of melanoma patients, can be very symptomatic and survival in this group may be prolonged. Regional chemotherapy with melphalan delivered by isolated limb perfusion (ILP) or isolated limb infusion (ILI) are effective treatment options which are generally well tolerated. ILI is a less invasive and simpler alternative to ILP. ILI is tolerated better than ILP, though is probably less effective. Complete response rates are 45- 69% for ILP and 23-44% for ILI. The limb is often warmed to lower temperatures in ILI compared to ILP and the limb becomes progressively more hypoxic and acidotic during ILI, each of these parameters potentially having an effect on outcome. ILP & ILI are used primarily as palliative options when excision of in-transit metastases is unfeasible but can be used as an adjunctive procedure to surgery, for other tumour types such as merkel cell carcinoma, and can be repeated if indicated. For ILI correction of melphalan dose for ideal body weight has been shown to substantially decrease the rates of severe local toxicity while maintaining complete response rates, but overall response rate is reduced. Combination treatment with tumour necrosis factor α has been used with variable outcomes and new combinations with buthionine sulfoximine and ADH-1 are being investigated.