EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Antidiabetogenic oligostilbenoids and 3-ethyl-4-phenyl-3,4-dihydroisocoumarins from the bark of Shorea roxburghii.
PMID 22209731
Abstract
A methanol extract of the bark of Shorea roxburghii (Dipterocarpaceae) was found to inhibit plasma glucose elevation in sucrose-loaded mice. From the extract, three new 3-ethyl-4-phenyl-3,4-dihydroisocoumarins, 1'S-dihydrophayomphenol A(2) (1) and phayomphenols B(1) (2) and B(2) (3), were isolated together with 24 known compounds including 20 stilbenoids and oligostilbenoids. The structures of 1-3 were determined on the basis of their spectroscopic properties as well as of chemical evidences. Among the isolates, (-)-hopeaphenol (6), hemsleyanol D (8), (+)-α-viniferin (15), and (-)-balanocarpol (18) showed inhibitory activity against plasma glucose elevation in sucrose-loaded rats at doses of 100-200mg/kg, p.o. To clarify the mode of action of the antihyperglycemic property, effects of these oligostilbenoids on gastric emptying in mice, those on glucose uptake in isolated intestinal tissues as well as inhibitory activities against rat intestinal α-glucosidase and rat lens aldose reductase were examined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
572691
3,4′-5-Trihydroxystilbene-3-β-D-glucopyranoside, 97%
C20H22O8