EMAIL THIS PAGE TO A FRIEND
Oncology reports
Upregulation of β2-microglobulin expression in progressive human oral squamous cell carcinoma.
PMID 22211248
Abstract
The aim of the present study was to investigate β2-microglobulin (β2-M) expression in normal oral mucosa and progressive oral squamous cell carcinoma (OSCC) and to assess the clinical significance of β2-microglobulin expression. The study included 10 cases of normal oral mucosa epithelium specimens, 55 cases of primary OSCC specimens, and 25 cases of OSCC metastasis specimens. Immunohistochemistry was used to determine β2-M expression, and its correlation with clinicopathological factors in progressive OSCC was evaluated. Immunohistochemistry showed that strong β2-M expression was significantly asscociated with tumor size (T3, T4 vs. T1, T2; P=0.001), positive node status (N positive vs. N negative; P=0.000) and advanced clinical stage (Ⅲ, Ⅳ vs. Ⅰ, Ⅱ, P=0.000) in primary OSCC lesions. Compared to primary OSCC lesions, the frequency of β2-M expression was significantly increased in metastatic OSCC lesions (P=0.02). In addition, in vitro results from Western blotting showed increased β2-M expression in the two OSCC lines studied. Therefore, we speculate that the up-regulation of β2-M expression may contribute to the oncogenesis of human oral mucosa, tumor invasion and metastasis.