EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of benzo[d]imidazole derivatives as potential anti-cancer agents.
PMID 22225635
Abstract
We herein report the synthesis, biological activity and structure-activity relationship of derivatives of 5,6-dichloro-1-β-D-ribofuranosylbenzimidazole and benzo[d]imidazole. A lead compound 6o demonstrates potent anti-proliferative activity and the ability to induce cancer cell apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D147206
5,6-Dimethylbenzimidazole, ≥99%
C9H10N2