EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Anion inhibition studies of an α-carbonic anhydrase from the living fossil Astrosclera willeyana.
PMID 22227210
Abstract
An α-carbonic anhydrase (CA, EC 4.2.1.1) isolated from the living fossil sponge Astrosclera willeyana, Astrosclerin, was investigated for its inhibition profile with simple inorganic anions, complex anions and other small molecules known to interact with these zinc enzymes. Astrosclerin is a catalytically highly efficient enzyme, and is inhibited in the low micromolar range by sulfamide, sulfamic acid, phenylboronic acid and phenylarsonic acid, and in the submillimolar range by a variety of anions including fluoride, chloride, cyanate, thiocyanate, cyanide, hydrogen sulfide, bisulfate, stannate, perosmate, divanadate, perrhenate, perruthenate, selenocyanide, trithiocarbonate, diethyldithiocarbamate and iminodisulfonate. Less efficient Astrosclerin inhibitors were sulfate, bromide, iodide, azide, bicarbonate, carbonate, tetraborate and perchlorate (K(I)s of 5.11-30.6mM) whereas tetrafluoroborate was not at all inhibitory. Because Astrosclerin is involved in calcification processes in vivo, its anion inhibition profile may be important for future studies designed to shed light on the physiologic functions of α-CAs in marine organisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36232 Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
243973
Sodium bisulfite, ACS reagent, mixture of NaHSO3 and Na2S2O5
799394
Sodium bisulfite, anhydrous, free-flowing, mixture of NaHSO3 and Na2S2O5, Redi-Dri, ACS reagent
13438
Sodium bisulfite solution, purum, ~40%
HNaO3S
S4547
Sodium bromide, BioXtra, ≥99.0%
BrNa
02119
Sodium bromide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
BrNa
460591
Sodium bromide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
BrNa
229881
Sodium bromide, ≥99.99% trace metals basis
BrNa
310506
Sodium bromide, ACS reagent, ≥99.0%
BrNa
220345
Sodium bromide, ReagentPlus®, ≥99%
BrNa
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
31432
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
07982
Sodium chloride physiological solution, BioUltra, tablet
52455
Sodium chloride physiological solution, BioUltra, tablet
71710
Sodium iodide, puriss. p.a., ≥99.0% (AT)
INa
383112
Sodium iodide, ACS reagent, ≥99.5%
INa
217638
Sodium iodide, ReagentPlus®, ≥99%
INa
03129
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
INa
237213
Sodium nitrite, ACS reagent, ≥97.0%
NNaO2
S2252
Sodium nitrite, ReagentPlus®, ≥99.0%
NNaO2
13447
Sodium nitrite, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 250, 99-100.5% (calc. to the dried substance)
NNaO2
799416
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
NNaO2
914
Sodium nitrite solution