EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Direct amination of α-substituted nitroacetates using di-tert-butyl azodicarboxylate catalyzed by Hatakeyama's catalyst β-ICD.
PMID 22231285
Abstract
We report the first example of catalytic asymmetric direct amination of α-monosubstituted nitroacetates using di-tert-butyl azodicarboxylate. The simple and easily available Hatakeyama's catalyst β-ICD 11 was found to be a highly enantioselective catalyst for this reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11625
Di-tert-butyl azodicarboxylate, purum, ≥98.0% (GC)
C10H18N2O4