EMAIL THIS PAGE TO A FRIEND
Nature immunology
The neuroimmune guidance cue netrin-1 promotes atherosclerosis by inhibiting the emigration of macrophages from plaques.
PMID 22231519
Abstract
Atherosclerotic plaque formation is fueled by the persistence of lipid-laden macrophages in the artery wall. The mechanisms by which these cells become trapped, thereby establishing chronic inflammation, remain unknown. Here we found that netrin-1, a neuroimmune guidance cue, was secreted by macrophages in human and mouse atheroma, where it inactivated the migration of macrophages toward chemokines linked to their egress from plaques. Acting via its receptor, UNC5b, netrin-1 inhibited the migration of macrophages directed by the chemokines CCL2 and CCL19, activation of the actin-remodeling GTPase Rac1 and actin polymerization. Targeted deletion of netrin-1 in macrophages resulted in much less atherosclerosis in mice deficient in the receptor for low-density lipoprotein and promoted the emigration of macrophages from plaques. Thus, netrin-1 promoted atherosclerosis by retaining macrophages in the artery wall. Our results establish a causative role for negative regulators of leukocyte migration in chronic inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HUTR001C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Housekeeping Control
HUTR002C
MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Empty Vector Control
HUTR003C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 1
HUTR004C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 2
HUTR00012 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARNTL
HUTR00018 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RORA
HUTR00022 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, STAT3
HUTR00023 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP53
HUTR00030 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TRIAP1
HUTR00036 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, MPP5
HUTR00041 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARFIP1
HUTR00043 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, DHX40
HUTR00046 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, LATS2
HUTR00047 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RBL2
HUTR00238 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ATP5G2
HUTR00247 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, NDST3
HUTR00256 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, WNK2
HUTR00264 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, EDF1
HUTR00276 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, XCR1
HUTR00286 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RAB40A
HUTR00297 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TTR
HUTR00307 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, IFI27L1
HUTR00317 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, AMDHD2
HUTR00331 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SHKBP1
HUTR00338 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCDC123
HUTR00345 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCT6B
HUTR00484 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP73
HUTR00491 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TLR1
HUTR00499 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SPATA5L1
HUTR00514 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CGB1