EMAIL THIS PAGE TO A FRIEND
Organic letters
Construction of cyclopentanol derivatives via three-component coupling of silyl glyoxylates, acetylides, and nitroalkenes.
PMID 22235777
Abstract
The three-component coupling of Mg acetylides, silyl glyoxylates, and nitroalkenes results in a highly diastereoselective Kuwajima-Reich/vinylogous Michael cascade that provides tetrasubstituted silyloxyallene products. The regio- and diastereoselectivity were studied using DFT calculations. These silyloxyallenes were converted to cyclopentenols and cyclopentitols via a unique Lewis acid assisted Henry cyclization. The alkene functionality present in the cyclopentanol products can be elaborated using diastereoselective ketohydroxylation reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C112208
Cyclopentanol, 99%
C5H10O