EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design and characterization of optimized adenosine A₂A/A₁ receptor antagonists for the treatment of Parkinson's disease.
PMID 22239465
Abstract
The design and characterization of two, dual adenosine A(2A)/A(1) receptor antagonists in several animal models of Parkinson's disease is described. Compound 1 was previously reported as a potential treatment for Parkinson's disease. Further characterization of 1 revealed that it was metabolized to reactive intermediates that caused the genotoxicity of 1 in the Ames and mouse lymphoma L51784 assays. The identification of the metabolites enabled the preparation of two optimized compounds 13 and 14 that were devoid of the metabolic liabilities associated with 1. Compounds 13 and 14 are potent dual A(2A)/A(1) receptor antagonists that have excellent activity, after oral administration, across a number of animal models of Parkinson's disease including mouse and rat models of haloperidol-induced catalepsy, mouse and rat models of reserpine-induced akinesia, and the rat 6-hydroxydopamine (6-OHDA) lesion model of drug-induced rotation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
512125
3-Pyridinylboronic acid, ≥95.0%
C5H6BNO2