EMAIL THIS PAGE TO A FRIEND
Proceedings of the Japan Academy. Series B, Physical and biological sciences
Peculiarity of methoxy group-substituted phenylhydrazones in Fischer indole synthesis.
PMID 22241067
Abstract
We found that the Fischer indole synthesis of ethyl pyruvate 2-methoxyphenylhydrazone (5) with HCl/EtOH gave an abnormal product, ethyl 6-chloroindole-2-carboxylate (7), as the main product, with a smaller amount of ethyl 7-methoxyindole-2-carboxylate (6) as the normal product. This abnormal reaction was the result of a cyclization on the side with the substituent (methoxy group) of a benzene ring on phenylhydrazone, which was not previously observed. In this initial investigation, we focused on 1) the application of the above-mentioned abnormal Fischer indole synthesis, 2) the details of this reaction of phenylhydrazone with other kinds of substituents, 3) the mechanism of the first step of the Fischer indole synthesis, 4) the abnormal reaction in methoxydiphenylhydrazones, and 5) a synthetic device to avoid an abnormal reaction. The results of these studies are summarized herein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48157 Indole solution, 2000 μg/mL in methanol, certified reference material