EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Complex activities of the human Bloom's syndrome helicase are encoded in a core region comprising the RecA and Zn-binding domains.
PMID 22253018
Abstract
Bloom's syndrome DNA helicase (BLM), a member of the RecQ family, is a key player in homologous recombination (HR)-based error-free DNA repair processes. During HR, BLM exerts various biochemical activities including single-stranded (ss) DNA translocation, separation and annealing of complementary DNA strands, disruption of complex DNA structures (e.g. displacement loops) and contributes to quality control of HR via clearance of Rad51 nucleoprotein filaments. We performed a quantitative mechanistic analysis of truncated BLM constructs that are shorter than the previously identified minimal functional module. Surprisingly, we found that a BLM construct comprising only the two conserved RecA domains and the Zn(2+)-binding domain (residues 642-1077) can efficiently perform all mentioned HR-related activities. The results demonstrate that the Zn(2+)-binding domain is necessary for functional interaction with DNA. We show that the extensions of this core, including the winged-helix domain and the strand separation hairpin identified therein in other RecQ-family helicases, are not required for mechanochemical activity per se and may instead play modulatory roles and mediate protein-protein interactions.