EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Triplet imaging of oxygen consumption during the contraction of a single smooth muscle cell (A7r5).
PMID 22259112
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5013
G 418 disulfate salt, potency: ≥450 μg per mg (dried basis)
C20H40N4O10 · 2H2SO4