EMAIL THIS PAGE TO A FRIEND
The FEBS journal
The kynurenine pathway and quinolinic acid: pivotal roles in HIV associated neurocognitive disorders.
PMID 22260426
Abstract
This brief review will first consider HIV associated neurocognitive disorder followed by the current understanding of its neuropathogenesis. Against this background the role of the kynurenine pathway will be detailed. Evidence both direct and indirect will be discussed for involvement of the kynurenine pathway at each step in the neuropathogenesis of HIV associated neurocognitive disorder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P63204
2,3-Pyridinedicarboxylic acid, 99%
C7H5NO4