EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Enantioseparation of 1-arylethanols via a supramolecular chiral host consisting of N-(2-naphthoyl)-L-aspartic acid and an achiral diamine.
PMID 22273775
Abstract
A supramolecular chiral host consisting of N-(2-naphthoyl)-L-aspartic acid (L-1) and meso-1,2-diphenylethylenediamine (2) is effective in enantioseparation of 1-arylethanols (up to 96% ee with 100% inclusion ratio). Here we report three different methods to prepare the inclusion crystals and discuss the chiral recognition mechanism on the basis of X-ray crystallography results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
458511
meso-1,2-Diphenylethylenediamine, 98%
C14H16N2