EMAIL THIS PAGE TO A FRIEND
Epilepsia
Blocking seizures with the diuretic bumetanide: promises and pitfalls.
PMID 22280471
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1158000
Bumetanide, European Pharmacopoeia (EP) Reference Standard
C17H20N2O5S