EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Kinetic resolution of secondary alcohols using amidine-based catalysts.
PMID 22283696
Abstract
Kinetic resolution of racemic alcohols has been traditionally achieved via enzymatic enantioselective esterification and ester hydrolysis. However, there has long been considerable interest in devising nonenzymatic alternative methods for this transformation. Amidine-based catalysts (ABCs), a new class of enantioselective acyl transfer catalysts developed in our group, have demonstrated, inter alia, high efficacy in the kinetic resolution of benzylic, allylic, and propargylic secondary alcohols and 2-substituted cycloalkanols, and thus provide a viable alternative to enzymes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900542
(2S)-2-Phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole, 95%
C16H14N2S