EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Concise synthesis of (+)-conduritol F and inositol analogues from naturally available (+)-proto-quercitol and their glucosidase inhibitory activity.
PMID 22290077
Abstract
An effective synthesis of (+)-conduritol F, (+)-chiro- and (+)-epi-inositols from naturally available (+)-proto-quercitol is described. This synthetic method provides a concise synthesis of cyclitols in enantiomerically pure form. Of the synthesized cyclitols, (+)-conduritol F potently inhibits type I α-glucosidase with an IC(50) value of 86.1 μM, which is five times greater than the standard antidiabetic drug, acarbose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8980
Acarbose, ≥95%
C25H43NO18